Learn More
Weber Scientific L-W STAIN FOR MILK SMEARS
SDP
Catalog No. NC0400202
Encompass
Canadian - Xylene formula. All available formulations conform to APHA, Standard Methods for the Examination of Dairy Products. The Canadian stain contains some red basic fuchsia dye which gives the smear a pinkish background.
Catalog No. NC0400202 Supplier Weber Scientific Supplier No. 200120
Canadian - Xylene formula. All available formulations conform to APHA, Standard Methods for the Examination of Dairy Products. The Canadian stain contains some red basic fuchsia dye which gives the smear a pinkish background.