Learn More
ATCC L Wnt-3A; Transfected; Mouse
Supplier:  ATCC CRL2647
Encompass
L Wnt-3A is a cell line exhibiting fibroblast-like morphology that was isolated from the areolar adipose tissue of a 100-day-old, male mouse. This cell line is presently the best source for the production of Wnt-3A conditioned medium. This cell line was deposited by R Nusse.
Catalog No. 50-238-3608