Learn More
Apexbio Technology LLC L-XYLOSE-100MG
SDP
Catalog No. 5000568313
Encompass_Preferred
L-Xylose (CAS No 609-06-3) is a naturally occurring pentose sugar commonly used in biomedical research It plays a vital role in studies related to carbohydrate metabolism and absorption in human and animal models L-Xylose is known to engage pathways associated with monosaccharide transport and absorption specifically targeting glucose transporters and related proteins Its common in vitro activity is observed in the nanomolar to low micromolar range making it effective in precision studies Typical applications include use in cell models for studying metabolic pathways and in animal studies to evaluate intestinal absorption efficiency Additionally it serves a crucial role in drug screening for therapies targeting metabolic disorders Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68313 Supplier Apexbio Technology LLC Supplier No. C7153100
L-Xylose (CAS No 609-06-3) is a naturally occurring pentose sugar commonly used in biomedical research It plays a vital role in studies related to carbohydrate metabolism and absorption in human and animal models L-Xylose is known to engage pathways associated with monosaccharide transport and absorption specifically targeting glucose transporters and related proteins Its common in vitro activity is observed in the nanomolar to low micromolar range making it effective in precision studies Typical applications include use in cell models for studying metabolic pathways and in animal studies to evaluate intestinal absorption efficiency Additionally it serves a crucial role in drug screening for therapies targeting metabolic disorders Experimental concentrations typically depend on the specific research design