Learn More
Medchemexpress LLC L-(-)-xylose-2-13C | 478506-63-7 | 99.0% | 151.12 | C5H10O5 (13C at 2-position) | 10 MG
SDP
Catalog No. 5000298816
Encompass_Preferred
L-Xylose-2-13C is the 13C-labeled form of L-xylose used as an isotopically labeled standard or tracer in analytical and synthetic applications. It is suitable for use as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and as a raw material for synthesis of bioactive molecules.
  • Isotopically labeled at the 2-position for precise tracer studies.
  • High purity suitable for analytical applications.
  • Available in small pack sizes for method development and standards.
  • Solid, white to off-white appearance for easy handling.
Catalog No. 50-002-98816 Supplier Medchemexpress LLC Supplier No. HY78139S110MG
L-Xylose-2-13C is the 13C-labeled form of L-xylose used as an isotopically labeled standard or tracer in analytical and synthetic applications. It is suitable for use as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS, and as a raw material for synthesis of bioactive molecules.
  • Isotopically labeled at the 2-position for precise tracer studies.
  • High purity suitable for analytical applications.
  • Available in small pack sizes for method development and standards.
  • Solid, white to off-white appearance for easy handling.