Learn More
Medchemexpress LLC L-Xylose-2-13C | 478506-63-7 | 99.0% | C₄¹³CH₁₀O₅ | 50 MG
SDP
Catalog No. 5000434854
Encompass_Preferred
L-Xylose-2-13C is a carbon-13 labeled form of L-Xylose, a rare and levorotatory sugar. This compound serves as a raw material for the synthesis of various drugs and bioactive molecules.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34854 Supplier Medchemexpress LLC Supplier No. HY78139S150MG
L-Xylose-2-13C is a carbon-13 labeled form of L-Xylose, a rare and levorotatory sugar. This compound serves as a raw material for the synthesis of various drugs and bioactive molecules.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS