Learn More
Medchemexpress LLC L-Xylose-2-13C | 478506-63-7 | >99.0% | C413CH10O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY78139S15MG
Encompass_Preferred
L-Xylose-2-13C is the 13C labeled L-Xylose, a rare sugar and the levorotatory form of Xylose. It serves as a valuable raw material for the synthesis of various drugs and bioactive molecules.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34877