Learn More
Medchemexpress LLC L-Xylose | 609-06-3 | 97.0% | C₅H₁₀O₅ | 10 G
SDP
Catalog No. 5000434926
Encompass_Preferred
L-Xylose (L-(-)-Xylose) is a rare sugar and the levorotatory form of Xylose. It has a molecular weight of 150.13 and appears as a white to off-white solid. It is soluble in DMSO at 100 mg/mL. This product serves as a raw material for the synthesis of various drugs and bioactive molecules.
  • Suitable for use as a raw material in drug synthesis
  • Levorotatory form of Xylose
  • White to off-white solid appearance
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-34926 Supplier Medchemexpress LLC Supplier No. HY7813910G
L-Xylose (L-(-)-Xylose) is a rare sugar and the levorotatory form of Xylose. It has a molecular weight of 150.13 and appears as a white to off-white solid. It is soluble in DMSO at 100 mg/mL. This product serves as a raw material for the synthesis of various drugs and bioactive molecules.
  • Suitable for use as a raw material in drug synthesis
  • Levorotatory form of Xylose
  • White to off-white solid appearance
  • Soluble in DMSO at 100 mg/mL