Learn More
Medchemexpress LLC L-Xylose (L-(-)-Xylose) | 609-06-3 | 97.0% | C5H10O5 | 50 G
SDP
Catalog No. 5000433243
Encompass_Preferred
L-Xylose, also known as L-(-)-Xylose, is a rare sugar and the levorotatory form of Xylose. This chemical compound serves as a raw material for the synthesis of various drugs and bioactive molecules. It has a molecular weight of 150.13 and typically appears as a white to off-white solid with a purity of 97.0%.
  • Rare sugar
  • Levorotatory form of xylose
  • Raw material for synthesis of drugs and bioactive molecules
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO at 100 mg/mL
  • Initial source from microorganisms, specifically Escherichia coli
Catalog No. 50-004-33243 Supplier Medchemexpress LLC Supplier No. HY7813950G
L-Xylose, also known as L-(-)-Xylose, is a rare sugar and the levorotatory form of Xylose. This chemical compound serves as a raw material for the synthesis of various drugs and bioactive molecules. It has a molecular weight of 150.13 and typically appears as a white to off-white solid with a purity of 97.0%.
  • Rare sugar
  • Levorotatory form of xylose
  • Raw material for synthesis of drugs and bioactive molecules
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO at 100 mg/mL
  • Initial source from microorganisms, specifically Escherichia coli