Learn More
Waters Corp Waters UPLC Columns Method Development Kit, C18, 1.7 μm/1.8 μm, 2.1 mm x 100 mm, 4 pk
Catalog No. 50791020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The L1 UPLC method development kit combines 1.7 μm BEH C18, Shield RP18, and 1.8 μm HSS C18 and T3 column chemistries for efficient method development. Utilizing UPLC Technology, this kit improves throughput, reduces costs and analysis time, and maintains separation integrity. This supports faster product market entry and enhances assay profitability.
  • Designed for reversed phase separation with C18 chemistry.
  • Features a wide pH range of 1 pH to 12 pH and withstands up to 1240 Bar (18000 psi) maximum pressure.
  • Includes endcapped spherical particles (1.7 μm and 1.8 μm) with low silanol activity.
  • Equipped with Parker-style endfittings, 130 Å pore size.
  • Optimized for UPLC and UHPLC systems as a method development kit.
  • Inner diameter of 2.1 mm and length of 100 mm.
  • Contains 4 units per package and includes eCord technology.
Catalog No. 50-791-020 Supplier Waters Corp Supplier No. 176001606
Add to Cart
Add to Cart
Request a Quote
The L1 UPLC method development kit combines 1.7 μm BEH C18, Shield RP18, and 1.8 μm HSS C18 and T3 column chemistries for efficient method development. Utilizing UPLC Technology, this kit improves throughput, reduces costs and analysis time, and maintains separation integrity. This supports faster product market entry and enhances assay profitability.
  • Designed for reversed phase separation with C18 chemistry.
  • Features a wide pH range of 1 pH to 12 pH and withstands up to 1240 Bar (18000 psi) maximum pressure.
  • Includes endcapped spherical particles (1.7 μm and 1.8 μm) with low silanol activity.
  • Equipped with Parker-style endfittings, 130 Å pore size.
  • Optimized for UPLC and UHPLC systems as a method development kit.
  • Inner diameter of 2.1 mm and length of 100 mm.
  • Contains 4 units per package and includes eCord technology.