Learn More
Beckman Coulter L10 10Um Nist Traceab Filter Paper [Ea]
Catalog No. NC2164257 Shop All Beckman Coulter Products
Encompass
L10 Standard, nominal 10μm, Latex Particle (NIST Traceable), 1 x 15 mL Product No:C72516 NIST Traceable. Microsphere. For use with the Multisizer COULTER COUNTER particle analyzer
This item is not returnable. View return policy
Catalog No. NC2164257 Supplier Beckman Coulter Supplier No. C72516
L10 Standard, nominal 10μm, Latex Particle (NIST Traceable), 1 x 15 mL Product No:C72516 NIST Traceable. Microsphere. For use with the Multisizer COULTER COUNTER particle analyzer