Learn More
Poltex L10 Wipe Holder
SDP
Catalog No. 22000328
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic Mount
VHB Tape
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-328 Keyhole Slots
22-000-330 VHB Tape
22-000-329 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-000-328 Supplier Poltex Supplier No. WYPL10W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Holds one box of L10 Utility Wipes. Super durable PETG construction.
Specifications
Dimensions (L x W x H) 5.75 x 5.2 x 10 in.
Material PETG
Mounting Keyhole Slots
Description Holds L10 Wypall Utility Wipes