Learn More
Invivogen L18-MDP, 1 mg
SDP
Catalog No. NC0189063
Encompass
Muramyl dipeptide (MDP) is the minimal bioactive peptidoglycan motif common to all bacteria, the essential structure required for adjuvant activity in vaccines. MDP has been shown to be recognized by NOD2, but not TLR2, nor TLR2/1 or TLR2/6 associations.
Catalog No. NC0189063 Supplier Invivogen Supplier No. TLRLLMDP
Muramyl dipeptide (MDP) is the minimal bioactive peptidoglycan motif common to all bacteria, the essential structure required for adjuvant activity in vaccines. MDP has been shown to be recognized by NOD2, but not TLR2, nor TLR2/1 or TLR2/6 associations.