Learn More
Aviva Systems Biology Corp L1CAM Antibody - C-terminal region (ARP63103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against L1CAM. It was validated on Western Blot. . Alternate names: CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAML1, S10, SPG1, NCAM-L1, N-CAM-L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-877