Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L1CAM Antibody Pair [HRP]
SDP

Catalog No. NB009932
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009932 5 Plates
NB009931 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB009932 Supplier R&D Systems™ Supplier No. NBP2794465PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 47 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Line Novus
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human L1CAM.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Gene ID (Entrez) 3897
Gene Symbol L1CAM
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 39 ng of recombinant L1CAM.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-L1CAM monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-L1CAM monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target L1CAM
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.