Learn More
Medchemexpress LLC L1CAM Mouse HEK293 100ug | 100ug
SDP
Catalog No. 5000187832
Encompass_Preferred
The L1CAM protein is a neural cell adhesion molecule that plays a key role in cell adhesion dynamics and transmembrane signaling during brain development It contributes to neuronal migration axonal growth fasciculation and synaptogenesis L1CAM Protein Mouse (HEK293 His) is the recombinant mouse-derived L1CAM protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87832 Supplier Medchemexpress LLC Supplier No. HYP77732100UG
The L1CAM protein is a neural cell adhesion molecule that plays a key role in cell adhesion dynamics and transmembrane signaling during brain development It contributes to neuronal migration axonal growth fasciculation and synaptogenesis L1CAM Protein Mouse (HEK293 His) is the recombinant mouse-derived L1CAM protein expressed by HEK293 with C-His labeled tag