Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci L1TD1 Peptide
SDP

Catalog No. 16594389
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-389 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-389 Supplier ProSci Supplier No. 6439P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

L1TD1 peptide is used for blocking the activity of L1TD1 antibody.

Specifications

Gene ID (Entrez) 54596
Species Human
Accession Number NP_061952
Common Name LINE-1 type transposase domain containing 1
Concentration 200 μg/mL
Content And Storage Store L1TD1 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol L1TD1
Quantity 0.05 mg
Sequence 17 amino acids near the carboxy terminus of human L1TD1.
Synonym ECAT11, ECAT11, LINE-1 type transposase domain-containing protein 1, ES cell-associated protein 11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.