Learn More
Medchemexpress LLC L2P4 TFA | 98.3% | 1961.83 (free base) | 5 MG
SDP
Catalog No. 5000425225
Encompass_Preferred
L2P4 TFA is a peptide-based and EBNA1 targeting fluorescent probe. It inhibits EBNA1 homodimer formation and selectively inhibits EBV+ tumor growth. L2P4 TFA exhibits cytotoxicity in EBV-positive C666-1 cells with an LC50 of 46.4 μM.
  • Peptide-based probe
  • EBNA1 targeting
  • Inhibits EBNA1 homodimer formation
  • Selectively inhibits EBV+ tumor growth
  • Exhibits cytotoxicity in EBV-positive C666-1 cells
  • Suitable for cancer targeted therapy and immunotherapy research
  • Used in peptide-drug conjugates research
Catalog No. 50-004-25225 Supplier Medchemexpress LLC Supplier No. HYP10772A5MG
L2P4 TFA is a peptide-based and EBNA1 targeting fluorescent probe. It inhibits EBNA1 homodimer formation and selectively inhibits EBV+ tumor growth. L2P4 TFA exhibits cytotoxicity in EBV-positive C666-1 cells with an LC50 of 46.4 μM.
  • Peptide-based probe
  • EBNA1 targeting
  • Inhibits EBNA1 homodimer formation
  • Selectively inhibits EBV+ tumor growth
  • Exhibits cytotoxicity in EBV-positive C666-1 cells
  • Suitable for cancer targeted therapy and immunotherapy research
  • Used in peptide-drug conjugates research