Learn More
Poltex L30 Wypall Holder - HB Tape
SDP
Catalog No. 01259157
Change view
Click to view available options
Dimensions (L x W x H):
5.75 x 6.25 x 10 in.
Catalog No. Dimensions (L x W x H)
01-259-157 5.75 x 6.25 x 10 in.
1 options
N/A
Catalog No. 01-259-157 Supplier Poltex Supplier No. WYPL30T
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A
Holds and dispenses wipes and offers multiple mounting options with durable, easy-to-clean construction.
  • Holds and dispenses Wypall wipes
  • Available for Wypall L10 or Wypall L30 wipes
  • Available in Wall Mount (screws and anchors included)
  • Available in HB Tape Mount (high-bond tape included)
  • Available in Magnet Mount (powerful magnets included)
  • Durable and cleanable PETG construction
  • Consumables not included
Specifications
Product Type Wipe Dispenser
Color Clear
Dimensions (L x W x H) 5.75 x 6.25 x 10 in.
Holds Labwipes, Wipes
Material PETG
Mounting VHB Tape
No. of Compartments 1
Product Style Open Top and Front
Edges Rounded
Includes High Bond Tape for Mounting
Quantity 1 Ea.
Sterility Non-sterile