Learn More
Cayman Chemical L3MBTL1 PosItIve CNTRL 800nmol
SDP
Catalog No. 502892680
Encompass_Preferred
A covalent inhibitor of PME-1 (IC50 0.6 UM) reduces the levels of methylated PP2A in HEK293T cells stably overexpressing PME-1 at 20 UM. AMZ30 inhibits t-BHP-induced decreases in U87MG and U-251MG cells at 210 UM
Catalog No. 50-289-2680 Supplier Cayman Chemical Supplier No. 601033800 NMOL
A covalent inhibitor of PME-1 (IC50 0.6 UM) reduces the levels of methylated PP2A in HEK293T cells stably overexpressing PME-1 at 20 UM. AMZ30 inhibits t-BHP-induced decreases in U87MG and U-251MG cells at 210 UM