Learn More
Aviva Systems Biology Corp L3MBTL2 Antibody - C-terminal region (ARP39670_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39670_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against L3MBTL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human L3MBTL2. Alternate names: H-l(3)mbt-l, L3MBTApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-57131