Learn More
Aviva Systems Biology Corp L3MBTL2 Antibody - N-terminal region (ARP30081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against L3MBTL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human L3MBTL2. Alternate names: H-l(3)mbt-l, L3MBTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54118