Learn More
Aviva Systems Biology Corp L3MBTL4 Antibody - N-terminal region (ARP36226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against L3MBTL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human L3MBTL4. Alternate names: HsT1031ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56077