Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex L40 Wypall Dispenser, Bulk Wipe
SDP

Catalog No. 22000378
Change view
Click to view available options
Mounting:
Keyhole Slots
Magnetic Mount
VHB Tape
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-378 Keyhole Slots
22-000-376 Magnetic Mount
22-000-377 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-000-378 Supplier Poltex Supplier No. WYPL40BKW
  • $123.10
    $107.65 / Each of 1

    Save $15.45

    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Holds and dispenses quarter fold Wypall 40 wipes.

Durable and cleanable PETG construction

Specifications

Color Clear
Length (Metric) 170 mm
Width (Metric) 180 mm
Height (Metric) 205 mm
Mounting Keyhole Slots
For Use With (Application) Wypal 40 quarter fold bulk wipes

consumables not included

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.