Learn More
Medchemexpress LLC L57 acetate | 99.01% | 2483.87 (free base) | 5 MG
SDP
Catalog No. 5000423786
Encompass_Preferred
L57 acetate is a Low-density lipoprotein receptor-related protein 1 (LRP1)-binding peptide that demonstrates high affinity to LRP1 with a Ki of 45 nM. It exhibits blood-brain barrier (BBB) permeability and plasma stability, making it suitable as a carrier for Central Nervous System (CNS) drug delivery.
  • High affinity to LRP1
  • Blood-brain barrier permeability
  • Plasma stability
  • Suitable for CNS drug delivery
Catalog No. 50-004-23786 Supplier Medchemexpress LLC Supplier No. HYP10502A5MG
L57 acetate is a Low-density lipoprotein receptor-related protein 1 (LRP1)-binding peptide that demonstrates high affinity to LRP1 with a Ki of 45 nM. It exhibits blood-brain barrier (BBB) permeability and plasma stability, making it suitable as a carrier for Central Nervous System (CNS) drug delivery.
  • High affinity to LRP1
  • Blood-brain barrier permeability
  • Plasma stability
  • Suitable for CNS drug delivery