Learn More
Chemglass Life Sciences #15 Quartz O-Ring Joint, 18.0mm Tube O.D., Clamp Size/28, 116 O-Ring Size
SDP
Catalog No. CGQ013505
Encompass_Preferred
#15 Quartz O-Ring Joint, 18.0mm Tube O.D., Clamp Size/28, 116 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.
Catalog No. CGQ013505 Supplier Chemglass Life Sciences Supplier No. CGQ013505
#15 Quartz O-Ring Joint, 18.0mm Tube O.D., Clamp Size/28, 116 O-Ring Size. Clear fused quartz o-ring joint. Mating surface is formed so that quartz joints can be connected directly to CG-124 borosilicate o-ring joints. For clamps see CG-150.