Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro L60N-1S Super Erecta 1 in. High Ledge for Wire Shelving

Catalog No. 13460150 Shop All Metro Products
Change view
Click to view available options
For Use With (Equipment):
Metro Super Erecta Shelving
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-460-150 Metro Super Erecta Shelving
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-460-150 Supplier Metro Supplier No. L60N1S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metro Super Erecta 1 in. High Ledges prevent items from falling or protruding.

  • For stationary or mobile installations.
  • Compatible with wire shelving only.
  • Stainless Steel Finish.

Specifications

Product Type Stackable Ledge for Wire Shelving
Includes Lab Table
With Casters No
Certifications/Compliance NSF Listed
For Use With (Application) Lab Table
Autoclavable Not Autoclavable
For Use With (Equipment) Metro Super Erecta Shelving
Color Silver
Warranty Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing.
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.