Learn More
TARGETMOL CHEMICALS INC L82-G17 5MG
SDP
Catalog No. 502578921
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. L82-G17 is an uncompetitive LigI-selective inhibitor in human cells with utility as a probe of the catalytic activity and cellular functions of LigI. purity: 99%
Catalog No. 50-257-8921 Supplier TARGETMOL CHEMICALS INC Supplier No. T99615MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. L82-G17 is an uncompetitive LigI-selective inhibitor in human cells with utility as a probe of the catalytic activity and cellular functions of LigI. purity: 99%