Learn More
Siemens Healthineers LA Controls (US)
Catalog No. 23044779 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-044-779 Low
23-044-780 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-779 Supplier Siemens Healthineers Supplier No. 10873569
Add to Cart
Add to Cart
This item is not returnable. View return policy
LA Controls contain positive control plasma for use in Lupus Anticoagulant clotting test assays.
LA Controls are positive controls for lupus anticoagulant clotting assays using LA 1 Screening and LA 2 Confirmation Reagents.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Control
For Use With (Application) Thrombophilia Testing
Level Low
Quantity 6 x 1 mL