Learn More
Takara Bio LA PCR KIT VER. 2.1 50 RXN
SDP
Catalog No. 50443983
Encompass
PCR Kit, Version 2.1; Qty.: 50 rxns; helps to optimize your long PCR and includes polymerase, buffer system, control template and primers
Catalog No. 50-443-983 Supplier Takara Bio Supplier No. RR013A
PCR Kit, Version 2.1; Qty.: 50 rxns; helps to optimize your long PCR and includes polymerase, buffer system, control template and primers