Learn More
Trajan Scientific La-Pha-Pack (A Thermo Brand), Vial Headspace 18mm Screw 20mL Radius Bottom 23x75mm, 100 Pack
Catalog No. NC1039619
Encompass
La-Pha-Pack (A Thermo Brand), Vial Headspace 18mm Screw 20mL Radius Bottom 23x75mm 100 Pack
Catalog No. NC1039619 Supplier Trajan Scientific Supplier No. 18091307
La-Pha-Pack (A Thermo Brand), Vial Headspace 18mm Screw 20mL Radius Bottom 23x75mm 100 Pack