Learn More
Braintree Scientific La Pias Ear Tags, Mouse, S/S, number 101-200
SDP
Supplier:  Braintree Scientific 56793400
Encompass
La Pias Ear Tags - Animal Identification Made Easy Specially suited for use with small lab and research animals (mice, rats and other rodents). The light weight easy-to-read tags allow for trouble-free identification. Ear tags are numbered from 1-100 (available up to 999) Available as Stainless Steel (non-colored) or in 5 Once applied, they cannot be easily removed Laser-etched number provides durability Material Numbered Tag: Aluminum Clasp: Stainless Steel Size Mouse: 3.5mm O.D. with 3mm PinRat: 3.5mm O.D. with 5mm Pin **Sold for research purposes only and not for human use**
Catalog No. 50-195-4135