Learn More
Teknova LA Plates with 1.6% Skim Milk and 2% Agar. 100mm Plates, 30mL Fill, 20 Plates/Sleeve, Sterile.
SDP
Catalog No. 502387966
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth ofbacteria. It is also known asLuria brothorLuria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains ofEscherichia coli.
Catalog No. 50-238-7966 Supplier Teknova Supplier No. M2055
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth ofbacteria. It is also known asLuria brothorLuria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains ofEscherichia coli.