Learn More
Takara Bio TaKaRa LA Taq® DNA Polymerase (Mg2+ free buffer), 125 units
SDP
Catalog No. 50443972
Encompass
Takara LA Taq is a mixture of Taq Polymerase with a proofreading polymerase. The mixture is optimized for amplification of long DNA templates (routinely 20 kb and up to 48 kb depending on template type) with high fidelity. This LA Taq polymerase mix is supplied with separate tubes of optimized buffer (Mg2+ free), MgCl2, and dNTPs. 125 units
Catalog No. 50-443-972 Supplier Takara Bio Supplier No. RR002A
Takara LA Taq is a mixture of Taq Polymerase with a proofreading polymerase. The mixture is optimized for amplification of long DNA templates (routinely 20 kb and up to 48 kb depending on template type) with high fidelity. This LA Taq polymerase mix is supplied with separate tubes of optimized buffer (Mg2+ free), MgCl2, and dNTPs. 125 units