Learn More
Takara Bio TaKaRa LA Taq® DNA Polymerase with GC Buffer, 125 units
SDP
Catalog No. 50443997
Encompass
Takara LA Taq DNA polymerase paired with two special GC-optimized Buffers. This system is designed to amplify templates with up to 73% GC content, and up to 20 kb in length routinely (extension up to 48 kb is possible on some templates). 125 units
Catalog No. 50-443-997 Supplier Takara Bio Supplier No. RR02AG
Takara LA Taq DNA polymerase paired with two special GC-optimized Buffers. This system is designed to amplify templates with up to 73% GC content, and up to 20 kb in length routinely (extension up to 48 kb is possible on some templates). 125 units