Learn More
Adam Equipment LAB 214i - Luna Analytical Balances - Capacity: 210g, Readability: 0.0001g
Catalog No. NC3964819
Encompass_Preferred
Elevate your laboratory's precision and efficiency with Luna Precision Balances, designed to meet the rigorous demands of labs, manufacturing, and production environments. Luna balances excel in tasks such as density determination, purity analysis, and quality control, offering unmatched durability and performance. The included hook facilitates below-balance weighing, expanding your measurement capabilities. Luna balances are equipped with USB and RS-232 interfaces, ensuring seamless communication with PCs and printers. This connectivity supports data tracking within Good Laboratory Practices (GLP) guidelines.
Catalog No. NC3964819 Supplier Adam Equipment Supplier No. LAB 214I
Elevate your laboratory's precision and efficiency with Luna Precision Balances, designed to meet the rigorous demands of labs, manufacturing, and production environments. Luna balances excel in tasks such as density determination, purity analysis, and quality control, offering unmatched durability and performance. The included hook facilitates below-balance weighing, expanding your measurement capabilities. Luna balances are equipped with USB and RS-232 interfaces, ensuring seamless communication with PCs and printers. This connectivity supports data tracking within Good Laboratory Practices (GLP) guidelines.