Learn More
Research Products International Corp Lab 45's Graduated Bottle, Glass, 137 x 267 mm, 2000 ml Capacity, 6 per Case
SDP
Catalog No. 50212469
Encompass
Multi-purpose graduated bottles designed for convenient storing or transporting of liquids. Constructed of heavy duty borosilicate glass for durability and chemical resistance. Bottles feature a wide 45mm screw thread finish, printed graduations and marking spot. Wide opening prevents fluid surge when pouring and rounded shoulders allow complete drainage of liquid. Supplied with white polypropylene cap and pour ring attached to bottle.
Catalog No. 50-212-469 Supplier Research Products International Corp Supplier No. 219931
Multi-purpose graduated bottles designed for convenient storing or transporting of liquids. Constructed of heavy duty borosilicate glass for durability and chemical resistance. Bottles feature a wide 45mm screw thread finish, printed graduations and marking spot. Wide opening prevents fluid surge when pouring and rounded shoulders allow complete drainage of liquid. Supplied with white polypropylene cap and pour ring attached to bottle.