Learn More
Lab-Aids™ ABO-Rh Combination Blood Typing Kit

Description
Includes
- 32 Combi-Slide guide
- One of each drop-controlled bottle blood-typing serum, Type Anti-A, Type Anti-B and Type Anti-Rh
- 32 spatulas for Anti-A serum (blue)
- 32 spatulas for Anti-B serum (yellow)
- 32 spatulas for Anti-Rh serum (red)
- 32 pre-saturated alcohol pads
- 32 sterile disposable blood lancets
- 32 student worksheets and guides
- Instruction manual with MSDS
This popular Lab-Aids product combines all the features of Basic Blood Typing and Rh Blood Typing kits with a specially designed Slide Guide™. Provides information on the history of the blood factors, hereditary determination, individual typing techniques, comparisons of blood types in world populations, and blood typing birth problems.
- Determine ABO blood type and Rh factor
- Correlate agglutination to antigen production
- Observe and study Rh mechanisms and relationships to transfusion and genetics
- Classroom size: 32 students
Specifications
Specifications
| Product Type | Abo-Rh Combination Blood Typing Kit |
| For Use With (Application) | Observe and study rh mechanisms and relationships to transfusion and genetics |
| Includes | 32 Combi slide guide, one of each drop controlled bottle Blood typing Serum, type Anti-A, type Anti-B and type Anti-Rh, 32 spatulas for Anti-A Serum (blue), 32 spatulas for Anti-B Serum (yellow), 32 spatulas for Anti-Rh Serum (red), 32 pre-saturated Alcohol pads, 32 sterile disposable Blood lancets, 32 student worksheets and guides, Instruction manual with MSDS |
| Class Size | 32 Students |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.