Learn More
Lab-Aids™ DNA Extraction and Isolation Kit

Description
Includes
- Four bottles cell lysis solution
- Two bottles DNA precipitation solution (keep ice cold until used; can be kept in a freezer)
- Eight plastic test tubes
- Eight plastic droppers
- Eight funnels
- Eight plastic sleeves
- Eight heavy-duty polybags
- Eight graduated cups
- One package cheesecloth squares
- One package wood splints
- 32 student worksheets and guides
- Instruction manual
Requires
Strawberries
A 'low-tech' hands-on solution to presenting some high-tech topics. Extract DNA from strawberries and examine some physical properties. A complete classroom kit that provides everything necessary (except strawberries) along with specially prepared cell lysating solutions and DNA precipitating.
This kit allows students to easily extract DNA by first crushing the strawberry, then lysing to release the cell components and finally precipitating and viewing the DNA after spooling the DNA on a splint. This kit contains step-by-step student instructions and teacher background information.
It is designed for a class of 8 teams of 4 and contains all the material to conduct 2 extractions. This is an excellent lab activity for all levels of teaching and for transferring complex concepts from the abstract into real observations. It allows students to visualize DNA and understand some of its properties. It illustrates the enormous length of the DNA molecule.
Order Info
Class size: 32 students working in teams of four.
Specifications
Specifications
| Product Type | DNA Extraction and Isolation Kit |
| For Use With (Application) | Extract dna from strawberries and understand some of its properties |
| Includes | Teacher's guide with MSDS, 32 student worksheets and guides, 8 Plastic droppers, 8 Plastic sleeves, 8 Plastic test tubes, 8 poly bags (heavy duty), 4 cell lysis solution, bottles, 2 DNA precipitation solution, bottles, 8 funnels, 8 graduated cups, cheesecloth squares, Wooden splints |
| Requires | Strawberries |
| Time Range | 50 min. |
| Class Size | 32 Students |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.