Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab-Aids™ Food Nutrient Analysis Kit
SCI_ED

Catalog No. S19340
Encompass_Preferred
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S19340 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19340 Supplier Lab-Aids™ Supplier No. 6A

Includes

  • 30 test tubes
  • Dropper-controlled bottles of sugar, starch, lipid, protein and vitamin C solutions
  • All necessary reagents
  • Wax pencil
  • 30 student worksheets
  • Instruction manual with Material Safety Data Sheets

Requires

Hot water bath (for test of simple sugars)

Test common foods for presence of nutrients, quantitatively determine Vitamin C in foods and standardize all tests.

Working in teams, students are asked to test for, and identify, five classes of nutrients: sugar, starch, lipid, protein, and Vitamin C. Once the tests have been completed and their observations have been made, students are then asked to draw conclusions about their results, the controls, and samples that showed no reaction. It is necessary to have a hot water bath available for the test of simple sugars. This kit has a limited shelf-life. Includes Teacher guide, student sheets, assessment questions and SDS.

Specifications

For Use With (Application) Test and identify five classes of nutrients like sugar, starch, lipid, protein, and vitamin c in foods
Includes Teacher's guide with MSDS, 30 Student Worksheets and Guides, Drop control bottle of Glucose, Vitamin C, Protein, Starch Suspension, Vegetable Oil, 2 Fat Testing Solution, 2 Protein Testing Solution A and B, 2 Sugar Testing Solution, 2 Vitamin C Testing Solution, 2 Bottles of Starch Testing solution, 30 Test tubes
Time Range One ∽50 minute class period
Class Size 30 Students
Product Type Food Nutrient Analysis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.