Learn More
Research Products International Corp Lab Alert Trpl 3 Channel Timer
SDP
Catalog No. 502979103
Encompass
Triple Timer/Clock with three channels is designed to count up and down with simultaneous display. Time-Out and 60 second audible Alarm Channels may be started independently or simultaneously. Additional features include memory recall and timing capacity of 20 hours. Includes one AAA battery.
Catalog No. 50-297-9103 Supplier Research Products International Corp Supplier No. 820030
Triple Timer/Clock with three channels is designed to count up and down with simultaneous display. Time-Out and 60 second audible Alarm Channels may be started independently or simultaneously. Additional features include memory recall and timing capacity of 20 hours. Includes one AAA battery.