Learn More
Research Products International Corp Lab Armor Beads, 2 Liters
SDP
Catalog No. 50550332
Encompass
Lab Armor Beads are a revolutionary modern day concept for replacing water in baths, ice in ice buckets and aluminum blocks in dry block incubators. These unique thermal technology beads have a broad temperature range of -80°C to 180°C. Lab Armor Beads can also be used to replace aluminum blocks in dry block heaters. Help to maintain a clean and green environment in the laboratory. Improves energy efficiency of standard water baths by over 50%.
Stays Clean - Protected from water-borne contamination
Stays Organized - No more water accidents
Saves Time - Always on and ready to use
Eco-Friendly - Highly recyclable beads, fewer harmful biocides and more energy efficient.
Catalog No. 50-550-332 Supplier Research Products International Corp Supplier No. 299102
Lab Armor Beads are a revolutionary modern day concept for replacing water in baths, ice in ice buckets and aluminum blocks in dry block incubators. These unique thermal technology beads have a broad temperature range of -80°C to 180°C. Lab Armor Beads can also be used to replace aluminum blocks in dry block heaters. Help to maintain a clean and green environment in the laboratory. Improves energy efficiency of standard water baths by over 50%.
Stays Clean - Protected from water-borne contamination
Stays Organized - No more water accidents
Saves Time - Always on and ready to use
Eco-Friendly - Highly recyclable beads, fewer harmful biocides and more energy efficient.