Learn More
Poltex Lab Bench Vertical Organizer
Click to view available options
Includes:
Organizer Only
Pipette Controller Bracker
Pipetter Bracket
Pipetter Bracket, Pipette Controller Bracket
Description
- Super durable 1/4 in. thick PETG construction
- Holds pipette tips, bottles, etc.
- See image for dimensions
- Choose from 3, 4, or 5 pocket
- Comes with optional accessories: Pipettor bracket or pipette controller bracket
- Rubber bumpons on base for no slip placement
Specifications
Specifications
| Total Positions | 4 Pockets |
| Includes | Pipetter Bracket |
| Material | PETG |