Learn More
Poltex Lab Bench Vertical Organizer
SDP
Catalog No. p-7224360
Change view
Click to view available options
Includes:
Organizer Only
Pipette Controller Bracker
Pipetter Bracket
Pipetter Bracket, Pipette Controller Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-588 Organizer Only
22-000-589 Pipette Controller Bracker
22-000-590 Pipetter Bracket
22-000-591 Pipetter Bracket, Pipette Controller Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-000-588 Supplier Poltex Supplier No. 4VTORG
Only null left
Add to Cart
Add to Cart
Free up valuable lab bench space by going up with this Lab Bench Vertical Organizer!
  • Super durable 1/4 in. thick PETG construction
  • Holds pipette tips, bottles, etc.
  • See image for dimensions
  • Choose from 3, 4, or 5 pocket
  • Comes with optional accessories: Pipettor bracket or pipette controller bracket
  • Rubber bumpons on base for no slip placement
Specifications
Includes Organizer Only
Total Positions 4 Pockets
Material PETG