Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Lab Bench Vertical Organizer with Accessories
SDP

Catalog No. 22000372
Change view
Click to view available options
Holds:
3 pocket
5 pocket
Includes:
4-position Pipetter Bracket
4-position Pipetter Bracket, Pipette Controller Bracket
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Includes
22-000-372 5 pocket 4-position Pipetter Bracket, Pipette Controller Bracket
22-000-369 3 pocket 4-position Pipetter Bracket
22-000-370 3 pocket 4-position Pipetter Bracket, Pipette Controller Bracket
22-000-371 5 pocket 4-position Pipetter Bracket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-000-372 Supplier Poltex Supplier No. 5VTORGPIPTRBRKBRK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Frees up valuable benchtop space by going vertical.

  • Bracket mounts to either side
  • Durable PETG Construction
  • Fits most major brands of tips and pipettes

Specifications

Includes 4-position Pipetter Bracket, Pipette Controller Bracket
Holds 5 pocket
Dimensions (D x W x H) 6 x 7.5 x 12.5 in.
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.