Learn More
Ace Glass, Inc. Lab bottle, Duran, 1000mL, plain GL 45, no cap or pour ring, cs/10
SDP
Catalog No. 50978378
Encompass
Chemical-resistant and stable 1000mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-978-378 Supplier Ace Glass, Inc. Supplier No. 553960
Chemical-resistant and stable 1000mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.