Learn More
Ace Glass, Inc. Lab bottle, Duran, 100mL, plain, pouring ring & PTFE 45 PBT cap, cs/10
SDP
Catalog No. 50971905
Encompass
Chemical-resistant and stable 100mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-905 Supplier Ace Glass, Inc. Supplier No. 553910
Chemical-resistant and stable 100mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.