Learn More
Ace Glass, Inc. Lab bottle, Duran, 10mL, plain, pouring ring & GL 25PP blue cap, cs/10
SDP
Catalog No. 50971920
Encompass
Chemical-resistant and stable 10mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-920 Supplier Ace Glass, Inc. Supplier No. 553979
Chemical-resistant and stable 10mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL25 PP cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/10.