Learn More
Ace Glass, Inc. Lab bottle, Duran, 10mL, plain GL 25, no cap or pour ring, cs/10
SDP
Catalog No. 50971911
Encompass
Chemical-resistant and stable 10mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL25 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.
Catalog No. 50-971-911 Supplier Ace Glass, Inc. Supplier No. 553949
Chemical-resistant and stable 10mL Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with GL25 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/10.