Learn More
Ace Glass, Inc. Lab bottle, Duran, 15L, amber, GL 45, no ring or cap, cs/1
SDP
Catalog No. 50978391
Encompass
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-978-391 Supplier Ace Glass, Inc. Supplier No. 5539266
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with GL45 thread. Does not include cap or pouring ring. Can be hot-air sterilized or autoclaved. Case/1.