Learn More
Ace Glass, Inc. Lab bottle, Duran, 15L, amber, pouring ring & PTFE 45 PBT cap, cs/1
SDP
Catalog No. 50971946
Encompass
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
Catalog No. 50-971-946 Supplier Ace Glass, Inc. Supplier No. 5539241
Chemical-resistant and stable 15L Duran laboratory bottles, fabricated from borosilicate glass and ambered. Supplied graduated with PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.